Learn More
Medchemexpress LLC Calcium disodium EDTA dihydrate | 6766-87-6 | 99.9% | 410.0226 Da | C10H16CaN2Na2O10 | 10g
SDP
Catalog No. 5000269532
Encompass_Preferred
Ca -EDTA disodium dihydrate (Calcium disodium EDTA dihydrate) is an orally active metal chelating reagent exhibits bactericidal activities against periodontal pathogens Aggregatibacter actinomycetemcomitans Prevotella intermedia and Porphyromonas gingivalis[1] Ca -EDTA disodium dihydrate is effective chelating antidotes for lead-and cadmium poisoning[2][3]
Catalog No. 50-002-69532 Supplier Medchemexpress LLC Supplier No. HYW74986710G
Ca -EDTA disodium dihydrate (Calcium disodium EDTA dihydrate) is an orally active metal chelating reagent exhibits bactericidal activities against periodontal pathogens Aggregatibacter actinomycetemcomitans Prevotella intermedia and Porphyromonas gingivalis[1] Ca -EDTA disodium dihydrate is effective chelating antidotes for lead-and cadmium poisoning[2][3]