Learn More
Medchemexpress LLC Calcium dobesilate | 20123-80-2 | 99.8% | 418.41 | 10 G
SDP
Catalog No. 5000439671
Encompass_Preferred
Calcium dobesilate is an orally active vascular protector that acts as an antioxidant, clearing hydroxyl free radicals and reducing oxidative stress and inflammation. It is studied for its potential in treating chronic venous diseases, diabetic retinopathy, and hemorrhoids.
  • Orally active vascular protector
  • Clears hydroxyl free radicals
  • Exhibits antioxidant activity
  • Reduces oxidative stress and inflammation
  • Decreases lipid peroxidation
  • Lowers MDA levels in red blood cells
  • Inhibits diabetes-induced increases in inflammatory markers (NF-κB, IL-6, IL-8, TNF-α, MCP-1)
  • Suppresses IL-1β-induced NF-kβ activation
  • Reduces TNF-α-induced upregulation of IL-6 and IL-18
Catalog No. 50-004-39671 Supplier Medchemexpress LLC Supplier No. HY11160310G
Calcium dobesilate is an orally active vascular protector that acts as an antioxidant, clearing hydroxyl free radicals and reducing oxidative stress and inflammation. It is studied for its potential in treating chronic venous diseases, diabetic retinopathy, and hemorrhoids.
  • Orally active vascular protector
  • Clears hydroxyl free radicals
  • Exhibits antioxidant activity
  • Reduces oxidative stress and inflammation
  • Decreases lipid peroxidation
  • Lowers MDA levels in red blood cells
  • Inhibits diabetes-induced increases in inflammatory markers (NF-κB, IL-6, IL-8, TNF-α, MCP-1)
  • Suppresses IL-1β-induced NF-kβ activation
  • Reduces TNF-α-induced upregulation of IL-6 and IL-18