Learn More
Medchemexpress LLC Calcium dobesilate | 20123-80-2 | 99.82% | 418.41 | 1 ML
SDP
Catalog No. 5000438872
Encompass_Preferred
Calcium dobesilate is an orally active vascular protector that can clear hydroxyl free radicals. It exhibits antioxidant activity, helping to reduce oxidative stress and inflammation. It is used for studying various conditions including chronic venous diseases, diabetic retinopathy, and hemorrhoids.
  • Orally active vascular protector
  • Clears hydroxyl free radicals with an IC50 of 1.1 pM
  • Exhibits antioxidant activity
  • Reduces oxidative stress and inflammation in retinas of diabetic mice
  • Used for studying chronic venous diseases
  • Used for studying diabetic retinopathy
  • Used for studying hemorrhoids
Catalog No. 50-004-38872 Supplier Medchemexpress LLC Supplier No. HY11160310MM/1ML
Calcium dobesilate is an orally active vascular protector that can clear hydroxyl free radicals. It exhibits antioxidant activity, helping to reduce oxidative stress and inflammation. It is used for studying various conditions including chronic venous diseases, diabetic retinopathy, and hemorrhoids.
  • Orally active vascular protector
  • Clears hydroxyl free radicals with an IC50 of 1.1 pM
  • Exhibits antioxidant activity
  • Reduces oxidative stress and inflammation in retinas of diabetic mice
  • Used for studying chronic venous diseases
  • Used for studying diabetic retinopathy
  • Used for studying hemorrhoids