Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Dobesilate Hydrate 98.0+%, TCI America™
SDP

Catalog No. C27891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27891G Supplier TCI America Supplier No. C27891G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 117552-78-0
Molecular Formula C12H12CaO11S2
Molecular Weight (g/mol) 436.415
MDL Number MFCD00867249
InChI Key JMTQBGBMQZPWCS-UHFFFAOYSA-L
Synonym Calcium 2,5-Dihydroxybenzenesulfonate, 2,5-Dihydroxybenzenesulfonic Acid Calcium Salt
PubChem CID 45358052
IUPAC Name calcium;2,5-dihydroxybenzenesulfonate;hydrate
SMILES C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.O.[Ca+2]

Specifications

Color White
Quantity 1 g
Formula Weight 418.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Dobesilate Hydrate
RTECSNumber DB6127000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.