Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices Fura-2 QBT Calcium Kit, Bulk, Ratiometric, 10 Vials

Catalog No. 5000014641
Encompass_Preferred

The Fura-2 QBT Calcium Kit (bulk) is a homogeneous, mix-and-read ratiometric assay that uses Fura-2 dye with extracellular masking technology to reduce wash steps and minimize assay variability. The bulk format supplies multiple vials for high-throughput screening on 96- and 384-well microplates.

  • Contains 10 vials of Component A
  • Each vial sufficient for approximately 10 microplates
  • Total kit coverage approximately 100 microplates (96- or 384-well)
  • Assay uses ratiometric Fura-2 dye, dual-excitation (340/380 nm), emission 510 nm
  • Compatible with fluorescence plate readers and FLIPR/FLIPR Tetra systems
  • Assay buffer (Component B) not included and must be obtained separately
  • Free of animal-derived components

Catalog No. 50-000-14641 Supplier Molecular Devices Supplier No. R8198
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Fura-2 QBT Calcium Kit (bulk) is a homogeneous, mix-and-read ratiometric assay that uses Fura-2 dye with extracellular masking technology to reduce wash steps and minimize assay variability. The bulk format supplies multiple vials for high-throughput screening on 96- and 384-well microplates.

  • Contains 10 vials of Component A
  • Each vial sufficient for approximately 10 microplates
  • Total kit coverage approximately 100 microplates (96- or 384-well)
  • Assay uses ratiometric Fura-2 dye, dual-excitation (340/380 nm), emission 510 nm
  • Compatible with fluorescence plate readers and FLIPR/FLIPR Tetra systems
  • Assay buffer (Component B) not included and must be obtained separately
  • Free of animal-derived components

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.