Learn More
Medchemexpress LLC Calcium fluoride | 7789-75-5 | MFCD00010907 | 98.8% | 78.07 | CaF2 | 25 G
SDP
Catalog No. 5000303977
Encompass_Preferred
Calcium fluoride (CaF2) is an inorganic solid supplied as a fine powder for laboratory and analytical use. It is commonly used as a source of fluoride and calcium in biochemical assays, materials research, and optical applications where a low-solubility fluoride is required. Documentation and handling guidance are provided for safe use and storage.
  • High purity solid suitable for research-grade applications.
  • Powder form facilitates dispersion and sample preparation.
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Chemical formula CaF2 and molecular weight 78.07 for easy reference.
  • Store sealed away from moisture; recommended cool storage.
Catalog No. 50-003-03977 Supplier Medchemexpress LLC Supplier No. 25GHYW019962
Calcium fluoride (CaF2) is an inorganic solid supplied as a fine powder for laboratory and analytical use. It is commonly used as a source of fluoride and calcium in biochemical assays, materials research, and optical applications where a low-solubility fluoride is required. Documentation and handling guidance are provided for safe use and storage.
  • High purity solid suitable for research-grade applications.
  • Powder form facilitates dispersion and sample preparation.
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Chemical formula CaF2 and molecular weight 78.07 for easy reference.
  • Store sealed away from moisture; recommended cool storage.