Learn More
Medchemexpress LLC Calcium fluoride | 7789-75-5 | MFCD00010907 | ≥99.0% | 78.07 | CaF2 | 500 G
SDP
Catalog No. 5000300088
Encompass_Preferred
Calcium fluoride (CaF2) is an inorganic, white crystalline solid supplied as a dry powder for laboratory use. It is commonly used as a source of fluoride or as an inert inorganic material in biochemical assays and research applications where low solubility and high chemical stability are desirable.
  • High purity (≥99.0%) for reliable experimental results.
  • Inorganic CaF2 composition with molecular weight 78.07.
  • Low solubility in water for stable performance in assays.
  • Available in multiple pack sizes, including 500 g, for flexible provisioning.
  • Documentation provided: data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-003-00088 Supplier Medchemexpress LLC Supplier No. HYW019962500G
Calcium fluoride (CaF2) is an inorganic, white crystalline solid supplied as a dry powder for laboratory use. It is commonly used as a source of fluoride or as an inert inorganic material in biochemical assays and research applications where low solubility and high chemical stability are desirable.
  • High purity (≥99.0%) for reliable experimental results.
  • Inorganic CaF2 composition with molecular weight 78.07.
  • Low solubility in water for stable performance in assays.
  • Available in multiple pack sizes, including 500 g, for flexible provisioning.
  • Documentation provided: data sheet, certificate of analysis, and safety data sheet.