Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calcium fluoride | 7789-75-5 | MFCD00010907 | ≥99.0% | 78.07 g/mol | CaF2 | 10 G
SDP

Catalog No. 5000301706
Encompass_Preferred

Calcium fluoride (CaF2) is an inorganic salt supplied as a white, crystalline powder for laboratory research. It is used as a reagent and biochemical assay material, notable for its chemical stability and low solubility. Review the product datasheet and SDS for specific handling, storage, and safety guidance before use.

  • High purity: ≥99.0%.
  • Chemical formula: CaF2.
  • Molecular weight: 78.07 g/mol.
  • Supplied as a powder for ease of weighing and handling.
  • Available in small to bulk pack sizes, including 10 g.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-003-01706 Supplier Medchemexpress LLC Supplier No. HYW01996210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calcium fluoride (CaF2) is an inorganic salt supplied as a white, crystalline powder for laboratory research. It is used as a reagent and biochemical assay material, notable for its chemical stability and low solubility. Review the product datasheet and SDS for specific handling, storage, and safety guidance before use.

  • High purity: ≥99.0%.
  • Chemical formula: CaF2.
  • Molecular weight: 78.07 g/mol.
  • Supplied as a powder for ease of weighing and handling.
  • Available in small to bulk pack sizes, including 10 g.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.