Learn More
Selleck Chemical LLC Calcium Folinate S2588-50mg
SDP
Supplier:  Selleck Chemical LLC S258850MG
Encompass_Preferred
Calcium Folinate is a calcium salt of folinic acid that is an adjuvant used in cancer chemotherapy Solutions are unstable and should be fresh-prepared
Catalog No. 50-311-0227