Learn More
Selleck Chemical LLC Calcium folinate (Leucovorin calcium) S5136-1g
SDP
Catalog No. 503112065
Encompass_Preferred
Calcium folinate (Leucovorin calcium) is a metabolically active reduced form of folic acid that is rapidly absorbed and extensively metabolized in the liver to other folic acid derivatives Solutions are unstable and should be fresh-prepared
Catalog No. 50-311-2065 Supplier Selleck Chemical LLC Supplier No. S51361G
Calcium folinate (Leucovorin calcium) is a metabolically active reduced form of folic acid that is rapidly absorbed and extensively metabolized in the liver to other folic acid derivatives Solutions are unstable and should be fresh-prepared