Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Calcium Free Salt Mixture, Modified

No Calcium in salt mixture.

$220.00 - $4131.00

Specifications

Product Type Animal Diet
Autoclavable Not Autoclavable
Disposable Single-use
Sterility Non-sterile
Storage Requirements Room Temperature
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP296020402
View Documents
MP Biomedicals
0296020402
2 kg
Each for $220.00
Only null left
Add to Cart
 
MP296020410
View Documents
MP Biomedicals
0296020410
10 kg
Each for $954.00
Only null left
Add to Cart
 
MP296020450
View Documents
MP Biomedicals
0296020450
50 kg
Each for $4,131.00
Only null left
Add to Cart
 
Description

Description

  • Salt mixture for multiple uses
  • No calcium in mixture
  • Modified to include essential trace minerals
  • This composition is high in phosphate
Specifications

Specifications

Animal Diet
Single-use
Room Temperature
0.3048 m
Not Autoclavable
Non-sterile
1 ft.
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.