Learn More
Apexbio Technology LLC Calcium Gluceptate 17140-60-2 10mM (in 1mL H2O)
SDP
Catalog No. 5000096196
Encompass_Preferred
Calcium Gluceptate (CAS 17140-60-2) is a calcium salt frequently utilized in biomedical research to modulate extracellular calcium concentrations and investigate calcium-dependent physiological processes including signal transduction muscle contraction and enzymatic functions By supplying bioavailable calcium ions it enables controlled studies of cellular calcium influx and related pathways Calcium Gluceptate is available in diverse purity grades suitable for a range of experimental protocols and is applicable in pharmacological assays tissue culture supplementation and studies of calcium homeostasis
Catalog No. 50-000-96196 Supplier Apexbio Technology LLC Supplier No. A83875.1
Calcium Gluceptate (CAS 17140-60-2) is a calcium salt frequently utilized in biomedical research to modulate extracellular calcium concentrations and investigate calcium-dependent physiological processes including signal transduction muscle contraction and enzymatic functions By supplying bioavailable calcium ions it enables controlled studies of cellular calcium influx and related pathways Calcium Gluceptate is available in diverse purity grades suitable for a range of experimental protocols and is applicable in pharmacological assays tissue culture supplementation and studies of calcium homeostasis