Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Gluconate 1-Hydrate, DPL

Catalog No. 502985652
Encompass
Catalog No. 50-298-5652 Supplier Thermo Scientific Production Chemicals and Services Supplier No. D503071003
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Molecular Weight (g/mol) 448.4 g/mol

Specifications

Boiling Point Undetermined
Quantity 25 kg
Percent Purity 98.5 to 102.0%
Grade Reagent Ph. Eur.
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.