Learn More
Apexbio Technology LLC Calcium gluconate 66905-23-5 25mg
SDP
Catalog No. 5000089678
Encompass_Preferred
Calcium gluconate is a calcium salt used to modulate cellular calcium dynamics It dissociates in solution to release calcium ions (Ca ) thereby influencing intracellular calcium concentrations and associated physiological processes Calcium gluconate exerts its biological activity primarily through altering calcium-dependent signaling pathways second messenger systems and enzyme activities It is frequently employed as a calcium supplement in research involving calcium deficiency-related disorders and studies targeting impaired calcium homeostasis Based on these properties calcium gluconate offers research potential in investigations of calcium signaling physiological responses related to calcium balance and therapeutic interventions addressing calcium deficiency
Catalog No. 50-000-89678 Supplier Apexbio Technology LLC Supplier No. C646125
Calcium gluconate is a calcium salt used to modulate cellular calcium dynamics It dissociates in solution to release calcium ions (Ca ) thereby influencing intracellular calcium concentrations and associated physiological processes Calcium gluconate exerts its biological activity primarily through altering calcium-dependent signaling pathways second messenger systems and enzyme activities It is frequently employed as a calcium supplement in research involving calcium deficiency-related disorders and studies targeting impaired calcium homeostasis Based on these properties calcium gluconate offers research potential in investigations of calcium signaling physiological responses related to calcium balance and therapeutic interventions addressing calcium deficiency