Learn More
Alkali Scientific Calcium Gluconate, Monohydrate, 250 G
SDP
Catalog No. 5000660743
Encompass_Preferred
Calcium Gluconate Monohydrate is a high-purity reagent provided in a 250g package, ideal for use in laboratory experiments involving calcium regulation or supplementation. It is commonly used in the preparation of buffered solutions, enzyme assays, and cell culture media that require precise calcium ion concentrations. Calcium Gluconate is also used in pharmaceutical applications, particularly in the development of oral and intravenous calcium supplements. Its versatility and high purity make it an essential reagent in both academic and industrial laboratories focusing on calcium-dependent processes in biochemical, clinical, and pharmaceutical research.
Catalog No. 50-006-60743 Supplier Alkali Scientific Supplier No. ALC002CP
Calcium Gluconate Monohydrate is a high-purity reagent provided in a 250g package, ideal for use in laboratory experiments involving calcium regulation or supplementation. It is commonly used in the preparation of buffered solutions, enzyme assays, and cell culture media that require precise calcium ion concentrations. Calcium Gluconate is also used in pharmaceutical applications, particularly in the development of oral and intravenous calcium supplements. Its versatility and high purity make it an essential reagent in both academic and industrial laboratories focusing on calcium-dependent processes in biochemical, clinical, and pharmaceutical research.