Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Gluconate, Monohydrate, USP, 98.5-102%, Spectrum™ Chemical
SDP

Catalog No. 18601306 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-306 2.5 kg Poly Pail
18-601-307 12 kg Poly Pail
18-601-308 50 kg Fiber Drum
18-601-305 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-306 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA1462.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

CA146, 66905-23-5, C12H22CaO14·H2O

Spectrum™ Chemical Calcium Gluconate, Monohydrate, USP is used as a nutrient. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 66905-23-5
Molecular Formula C12H24CaO15
Molecular Weight (g/mol) 448.39
InChI Key XLNFVCRGJZBQGX-UHFFFAOYNA-L
IUPAC Name calcium bis(2,3,4,5,6-pentahydroxyhexanoate) hydrate
SMILES O.[Ca++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 2.5 kg
Percent Purity 98.5 to 102%
Grade USP
Loss on Drying 2%
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.