Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Gluconate, Monohydrate, USP, 98.5-102%, Spectrum™ Chemical
SDP

Catalog No. 18601308 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-308 50 kg Poly Bottle
18-601-306 2.5 kg Poly Bottle
18-601-307 12 kg Poly Bottle
18-601-305 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-308 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA14650KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CA146, 66905-23-5, C12H22CaO14·H2O

Spectrum™ Chemical Calcium Gluconate, Monohydrate, USP is used as a nutrient. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 66905-23-5
Molecular Formula C12H24CaO15
Molecular Weight (g/mol) 448.39
InChI Key XLNFVCRGJZBQGX-UHFFFAOYNA-L
IUPAC Name calcium bis(2,3,4,5,6-pentahydroxyhexanoate) hydrate
SMILES O.[Ca++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 98.5 to 102%
Grade USP
Loss on Drying 2%
Packaging Poly Bottle
Chemical Name or Material Calcium Gluconate, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.