Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Glycolate 97.0+%, TCI America™
SDP

Catalog No. G020025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G020025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G020025G Supplier TCI America Supplier No. G020025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 996-23-6
Molecular Formula C4H8CaO6
Molecular Weight (g/mol) 192.18
MDL Number MFCD00067202
InChI Key UPYDGZJYILCHSK-UHFFFAOYSA-N
Synonym Glycolic Acid Calcium Salt, Hydroxyacetic Acid Calcium Salt
PubChem CID 131888598
IUPAC Name calcium;2-hydroxyacetic acid
SMILES C(C(=O)O)O.C(C(=O)O)O.[Ca]

Specifications

Color White
Quantity 25 g
Formula Weight 190.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Glycolate
RTECSNumber MC5290000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.