Learn More
Medchemexpress LLC Calcium green-5N (hexapotassium) | 153130-66-6 | 1191.10 | 500 UG
SDP
Catalog No. 5000431150
Encompass_Preferred
Calcium green-5N hexapotassium is a cell-impermeant Ca²⁺-sensitive fluorescent probe. It shows enhancement in fluorescence intensity on binding Ca²⁺ and can monitor free Ca²⁺ in neurons.
  • Cell-impermeant Ca²⁺-sensitive fluorescent probe
  • Shows enhancement in fluorescence intensity on binding Ca²⁺
  • Monitors free Ca²⁺ in neurons
Catalog No. 50-004-31150 Supplier Medchemexpress LLC Supplier No. HYD1631A500UG
Calcium green-5N hexapotassium is a cell-impermeant Ca²⁺-sensitive fluorescent probe. It shows enhancement in fluorescence intensity on binding Ca²⁺ and can monitor free Ca²⁺ in neurons.
  • Cell-impermeant Ca²⁺-sensitive fluorescent probe
  • Shows enhancement in fluorescence intensity on binding Ca²⁺
  • Monitors free Ca²⁺ in neurons