Learn More
Aqua Solutions CALCIUM HARDNESS 100G POWDER
SDP
Catalog No. NC9739249
Encompass_Preferred
Calcium Hardness Indicator Powder, 0.2% Calcon In Sodium Chloride (100grams)
Catalog No. NC9739249 Supplier Aqua Solutions Supplier No. 1700100G
Calcium Hardness Indicator Powder, 0.2% Calcon In Sodium Chloride (100grams)