Learn More
Hach™ Calcium Hardness Reagent Set, for Digital Titrator
Catalog No. 501996020 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Range:
10 to 160mg/L CaCO3
Catalog No. Range
50-199-6020 10 to 160mg/L CaCO3
1 options
Catalog No. 50-199-6020 Supplier Hach™ Supplier No. 2447200
Reagent set for the determination of low range Calcium Hardness by the EDTA Digital Titrator Titration Method. Set contains CalVer™ 2 Calcium Indicator Powder Pillows, Potassium Hydroxide Solution (8N) and 0.0800M (EDTA) Digital Titrator Cartridge for approximately 40-100 tests.
  • Accurate results when and where you need them
  • Fast and Flexible, with no cleaning or rinsing burets
  • Versatile with interchangeable cartridges
  • Economical, concentrated titrants in each cartridge
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Low range hardness (calcium) determination
For Use With (Equipment) Digital titrator
Includes CalVer™ 2 Calcium Indicator Powder Pillows, Potassium Hydroxide Solution (8N), 0.0800M (EDTA) Digital Titrator Cartridge
No. of Tests 40 to 100
Quantity 1 Set
Range 10 to 160mg/L CaCO3
Test Method EDTA titration
Type Reagent set