Learn More
Medchemexpress LLC Calcium hydroxide | 1305-62-0 | 95% | 100 G
SDP
Catalog No. 5000411599
Encompass_Preferred
Calcium hydroxide, ACS, 95% exhibits high bactericidal activity and high pH. It has the ability to induce hard tissue formation and can dissociate into calcium (Ca) and hydroxyl ions (OH). This product also shows potential for intra-canal medicament research.
  • High bactericidal activity
  • High pH
  • Ability to induce hard tissue formation
  • Dissociates into calcium (Ca) and hydroxyl ions (OH)
  • Potential for intra-canal medicament research
Catalog No. 50-004-11599 Supplier Medchemexpress LLC Supplier No. HYB2217D100G
Calcium hydroxide, ACS, 95% exhibits high bactericidal activity and high pH. It has the ability to induce hard tissue formation and can dissociate into calcium (Ca) and hydroxyl ions (OH). This product also shows potential for intra-canal medicament research.
  • High bactericidal activity
  • High pH
  • Ability to induce hard tissue formation
  • Dissociates into calcium (Ca) and hydroxyl ions (OH)
  • Potential for intra-canal medicament research