Learn More
Medchemexpress LLC Calcium hydroxide | 1305-62-0 | 95% | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYB2217
Encompass_Preferred
Calcium hydroxide, ACS, 95% has high bactericidal activity and high pH. It can induce hard tissue formation and dissociate into calcium (Ca) and hydroxyl ions (OH). It has the potential for intra-canal medicament research.
  • High bactericidal activity and high pH
  • Ability to induce hard tissue formation
  • Can dissociate into calcium (Ca) and hydroxyl ions (OH)
  • Potential for intra-canal medicament research
Catalog No. 50-004-14479