Learn More
Medchemexpress LLC Calcium hydroxide, ACS | 1305-62-0 | MFCD00010901 | 95% | 50 G
SDP
Catalog No. 5000409655
Encompass_Preferred
Calcium hydroxide, ACS, 95% exhibits high bactericidal activity and high pH. It has the ability to induce hard tissue formation and can dissociate into calcium (Ca) and hydroxyl ions (OH). This product has potential for intra-canal medicament research.
  • High bactericidal activity and high pH.
  • Ability to induce hard tissue formation.
  • Dissociates into calcium (Ca) and hydroxyl ions (OH).
  • Potential for intra-canal medicament research.
Catalog No. 50-004-09655 Supplier Medchemexpress LLC Supplier No. HYB2217D50G
Calcium hydroxide, ACS, 95% exhibits high bactericidal activity and high pH. It has the ability to induce hard tissue formation and can dissociate into calcium (Ca) and hydroxyl ions (OH). This product has potential for intra-canal medicament research.
  • High bactericidal activity and high pH.
  • Ability to induce hard tissue formation.
  • Dissociates into calcium (Ca) and hydroxyl ions (OH).
  • Potential for intra-canal medicament research.