Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Hypochlorite (Granular/Certified), Fisher Chemical

Catalog No. C100500 Shop All Fisherbrand Products
Change view
Click to view available options
Glass Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
C100-500 Glass Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C100-500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Specifications

Melting Point 100°C
Color White
CAS 7778-54-3
pH 11.4
Physical Form Solid
Quantity 500 g
Molecular Formula CaCl2O2
Linear Formula Ca(OCl)2
MDL Number MFCD00010900
Synonym calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor
InChI Key ZKQDCIXGCQPQNV-UHFFFAOYSA-N
SMILES [O-]Cl.[O-]Cl.[Ca+2]
IUPAC Name calcium;dihypochlorite
Molecular Weight (g/mol) 142.976
PubChem CID 24504
Formula Weight 142.98
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Density 2.350g/cm³
Chemical Name or Material Calcium Hypochlorite
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Harmful if swallowed. Contact with acids liberates toxic gas. Very toxic to aquatic organisms. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation DOT Class 5.1, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.