Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium(II) Bis(nonafluorobutanesulfonyl)imide 95.0+%, TCI America™
SDP

Catalog No. C34471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34471G 1 g
C34475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34471G Supplier TCI America Supplier No. C34471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 689282-13-1
Molecular Formula C16CaF36N2O8S4
Molecular Weight (g/mol) 1200.443
InChI Key WGPFQGXTGKSRPV-UHFFFAOYSA-N
Synonym Bis(nonafluorobutanesulfonyl)imide Calcium(II) Salt
PubChem CID 134128889
IUPAC Name calcium;bis(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)azanide
SMILES C(C(C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F.C(C(C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F.[Ca+2]

Specifications

Color White
Quantity 1 g
UN Number 1759
Formula Weight 1200.44
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium(II) Bis(nonafluorobutanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.