Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium iodate monohydrate, 99+%

Catalog No. AA4013836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4013836 500 g
AA4013822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4013836 Supplier Thermo Scientific Chemicals Supplier No. 04013836
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10031-32-0 | Ca(IO3)2·H2O

Calcium iodate monohydrate is used as oxidant added to lotions and ointments, acts as antiseptic and deodorant. It is used in the manufacture of disinfectants and iodine supplement in animal feed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-32-0
Molecular Formula Ca(IO3)2·H2O
MDL Number MFCD00036265

Specifications

Color White
Physical Form Powder
Quantity 500 g
Appearance Off-white powder
Assay Percent Range >99%
Solubility Information Soluble in nitric acid and slightly soluble in water. Insoluble in ethanol.
Formula Weight 407.90 (389.88 Anhydrous)
Chemical Name or Material Calcium iodate monohydrate
Hazard Category H272-H315-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 232-191-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.