Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium iodide hydrate

Catalog No. AA4013636
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4013636 500 g
AA4013622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4013636 Supplier Thermo Scientific Chemicals Supplier No. 04013636
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 71626-98-7 | CaI2 | 293.89 g/mol

Calcium iodide finds use in the preparation of complex used in polymerization and hydroamination as well as serving as a matrix for luminescent materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71626-98-7
Molecular Formula CaI2
Molecular Weight (g/mol) 293.89
MDL Number MFCD00149622
InChI Key UNMYWSMUMWPJLR-UHFFFAOYSA-L
IUPAC Name calcium diiodide
SMILES [Ca++].[I-].[I-]

Specifications

Melting Point 42°C (decomposition)
Color White to Gray
Physical Form Lump
Quantity 500 g
Sensitivity Hygroscopic
Density 2.55 g/mL
Merck Index 14,1677
Solubility Information Freely soluble in water and alcohol
Formula Weight 293.89 (Anhydrous)
Chemical Name or Material Calcium iodide hydrate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 233-276-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.