Learn More
Cell Signaling Technology Calcium Ion Regulation Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502730670
Encompass
The Calcium Ion Regulation Antibody Sampler Kit provides an economical way to investigate the regulation of calcium ions within the cell. This kit contains a selection of primary and secondary antibodies to analyze proteins involved in calcium ion regulation, including sarcoplasmic and endoplasmic reticulum Ca2+ ATPases (SERCA) and phospholamban (PLN). Each kit provides enough primary and secondary antibodies to perform two western blot experiments per primary antibody.
  • Includes primary and secondary antibodies for western blotting
  • Allows for the analysis of multiple targets involved in calcium ion regulation
  • Provides sufficient materials for two western blot experiments per primary antibody
  • Validated for use in research applications
Catalog No. 50-273-0670 Supplier Cell Signaling Technology Supplier No. 8575T
The Calcium Ion Regulation Antibody Sampler Kit provides an economical way to investigate the regulation of calcium ions within the cell. This kit contains a selection of primary and secondary antibodies to analyze proteins involved in calcium ion regulation, including sarcoplasmic and endoplasmic reticulum Ca2+ ATPases (SERCA) and phospholamban (PLN). Each kit provides enough primary and secondary antibodies to perform two western blot experiments per primary antibody.
  • Includes primary and secondary antibodies for western blotting
  • Allows for the analysis of multiple targets involved in calcium ion regulation
  • Provides sufficient materials for two western blot experiments per primary antibody
  • Validated for use in research applications