Learn More
Novus Biologicals™ Calcium Ionophore

Description
Application Notes: Ionophore is highly selective for Ca2+. Potentiates responses to NMDA but not quisqualate. In cell culture, stimulates nitric oxide production by calmodulin-dependent constitutive nitric oxide synthase.
Solubility: Ethanol: 5 mg/mL (with heating. Solutions are stable for at least 3 months at -20°C when protected from light and water contamination.) DMSO: 50 mg/mL (with heating or sonication. Solutions are stable for at least 3 months at -20°C when protected from light and water contamination.) Aqueous solutions can be made by diluting DMSO solutions in H2O.
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark. |
| Quantity | 1 mg |
| Target | Calcium Ionophore |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.