Learn More
Apexbio Technology LLC Calcium Ionophore I 58801-34-6 5mg
SDP
Catalog No. 5000088647
Encompass_Preferred
Calcium Ionophore I (also known as CA 1001 or ETH 1001 CAS 58801-34-6) is a neutral small molecule designed to selectively transport Ca2 ions across biological membranes By functioning as a Ca2 -selective ion carrier it modulates membrane ion permeability enabling controlled influx of calcium into cells such as human erythrocytes Studies show ETH 1001 increases intracellular Ca2 levels when external calcium is present and promotes Ca2 flux in loaded vesicles derived from red blood cells This compound is also employed in the fabrication of Ca2 -selective microelectrodes for membrane potential and ion concentration measurements in physiological and neurobiological research
Catalog No. 50-000-88647 Supplier Apexbio Technology LLC Supplier No. B60655
Calcium Ionophore I (also known as CA 1001 or ETH 1001 CAS 58801-34-6) is a neutral small molecule designed to selectively transport Ca2 ions across biological membranes By functioning as a Ca2 -selective ion carrier it modulates membrane ion permeability enabling controlled influx of calcium into cells such as human erythrocytes Studies show ETH 1001 increases intracellular Ca2 levels when external calcium is present and promotes Ca2 flux in loaded vesicles derived from red blood cells This compound is also employed in the fabrication of Ca2 -selective microelectrodes for membrane potential and ion concentration measurements in physiological and neurobiological research