Learn More
Medchemexpress LLC Calcium ionophore I (ETH 1001) | 58801-34-6 | MFCD00042943 | 99.6% | 684.99 | 1 MG
SDP
Catalog No. 5000351601
Encompass_Preferred
Calcium ionophore I (ETH 1001) is a selective Ca2+ ionophore for biological membranes. It can be utilized in Ca2+-selective microelectrodes for quantitative intracellular measurements of resting Ca2+-activities and slowly changing Ca2+-levels. This product is intended for research use only and promotes Ca2+ flux across RBC membranes without affecting Mg2+ movement.
  • Selective Ca2+ ionophore for biological membranes.
  • Can be used in Ca2+-selective microelectrodes for quantitative intracellular measurements of resting Ca2+-activities and slowly changing Ca2+-levels.
  • Promotes Ca2+ flux across RBC membranes without affecting Mg2+ movement.
Catalog No. 50-003-51601 Supplier Medchemexpress LLC Supplier No. HY1364601MG
Calcium ionophore I (ETH 1001) is a selective Ca2+ ionophore for biological membranes. It can be utilized in Ca2+-selective microelectrodes for quantitative intracellular measurements of resting Ca2+-activities and slowly changing Ca2+-levels. This product is intended for research use only and promotes Ca2+ flux across RBC membranes without affecting Mg2+ movement.
  • Selective Ca2+ ionophore for biological membranes.
  • Can be used in Ca2+-selective microelectrodes for quantitative intracellular measurements of resting Ca2+-activities and slowly changing Ca2+-levels.
  • Promotes Ca2+ flux across RBC membranes without affecting Mg2+ movement.