Learn More
TARGETMOL CHEMICALS INC Calcium ionophore II 25MG
SDP
Catalog No. 502880457
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Calcium ionophore II (N,N,N,N-Tetracyclohexyl-3-oxapentanediamide) is a lipophilic ionophore utilized in preparing calcium ion-selective electrodes. Purity 98.56%
Catalog No. 50-288-0457 Supplier TARGETMOL CHEMICALS INC Supplier No. T4085725MG
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Calcium ionophore II (N,N,N,N-Tetracyclohexyl-3-oxapentanediamide) is a lipophilic ionophore utilized in preparing calcium ion-selective electrodes. Purity 98.56%