Learn More
Cayman Chemical CalcIum Ionphore I 10mg
SDP
Catalog No. 502967434
Encompass_Preferred
A Ca2+-selective ionophore for biological membranes that can be used in microelectrodes for the determination of resting Ca2+-activities and of slowly changing Ca2+ levels
Catalog No. 50-296-7434 Supplier Cayman Chemical Supplier No. 1740710 MG
A Ca2+-selective ionophore for biological membranes that can be used in microelectrodes for the determination of resting Ca2+-activities and of slowly changing Ca2+ levels