Learn More
Cayman Chemical CalcIum Ionphore I 25mg
SDP
Supplier:  Cayman Chemical 1740725 MG
Encompass_Preferred
A Ca2+-selective ionophore for biological membranes that can be used in microelectrodes for the determination of resting Ca2+-activities and of slowly changing Ca2+ levels
Catalog No. 50-296-8159