Learn More
Cayman Chemical CalcIum Ionphore II 10mg
SDP
Catalog No. 502916809
Encompass_Preferred
A calcium ionophore; selective for calcium over strontium, manganese, cobalt, nickel, zinc, lanthanum, sodium, potassium, and magnesium; binds to calcium in a 1:3 (ligand:ion) ratio; induces calcium transport across artificial membranes and into rat liver mitochondria; induces sea urchin egg activation at 100 pM; has been used in the preparation of calcium-selective electrodes
Catalog No. 50-291-6809 Supplier Cayman Chemical Supplier No. 3770710 MG
A calcium ionophore; selective for calcium over strontium, manganese, cobalt, nickel, zinc, lanthanum, sodium, potassium, and magnesium; binds to calcium in a 1:3 (ligand:ion) ratio; induces calcium transport across artificial membranes and into rat liver mitochondria; induces sea urchin egg activation at 100 pM; has been used in the preparation of calcium-selective electrodes