Learn More
Cayman Chemical Calcium Ionophore II, Purity: ≥98%, CAS: 74267-27-9, 50 mg
SDP
Catalog No. 502918212
Encompass_Preferred
A calcium ionophore; selective for calcium over strontium, manganese, cobalt, nickel, zinc, lanthanum, sodium, potassium, and magnesium; binds to calcium in a 1:3 (ligand:ion) ratio; induces calcium transport across artificial membranes and into rat liver mitochondria; induces sea urchin egg activation at 100 pM; has been used in the preparation of calcium-selective electrodes
Catalog No. 50-291-8212 Supplier Cayman Chemical Supplier No. 3770750 MG
A calcium ionophore; selective for calcium over strontium, manganese, cobalt, nickel, zinc, lanthanum, sodium, potassium, and magnesium; binds to calcium in a 1:3 (ligand:ion) ratio; induces calcium transport across artificial membranes and into rat liver mitochondria; induces sea urchin egg activation at 100 pM; has been used in the preparation of calcium-selective electrodes