Learn More
Calcium L-Threonate, MilliporeSigma™ Supelco™
Catalog No. 111003787 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
Catalog No. Quantity
11-100-3787 500 mg
1 options
Catalog No. 11-100-3787 Supplier MilliporeSigma Supelco Supplier No. 1086877500MG
Add to Cart
Add to Cart
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY
Specifications
Purity Grade Notes These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Type USP Reference Standard
Quantity 500 mg
Grade Pharmaceutical Primary Standard