Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Lactate Pentahydrate (USP), Fisher Chemical

Catalog No. c110500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C110-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. C110-500
This item is not returnable. View return policy

Chemical Identifiers

CAS 5743-47-5
Molecular Formula C6H20CaO11
Molecular Weight (g/mol) 308.29
MDL Number MFCD00287281 (5H2O)
InChI Key HPVJXNNKHRNBOY-UHFFFAOYNA-L
Synonym calcium lactate pentahydrate, calcium 2-hydroxypropanoate pentahydrate, calcium lactate, anhydrous, c6h10cao6, calcium pentahydrate dilactate, calcii lactas, l-calcium lactate, l-?calcium?lactate, calcium lactate tn, lactic acid calcium salt
PubChem CID 165341
IUPAC Name calcium bis(2-hydroxypropanoate) pentahydrate
SMILES O.O.O.O.O.[Ca++].CC(O)C([O-])=O.CC(O)C([O-])=O

Specifications

Acidity Pass Test (USP)
Melting Point 240°C
Color White
Loss on Drying 22.0 to 27.0%
Assay Percent Range 98.0 to 101.0 %
Packaging Glass Bottle
Linear Formula Ca(CH3CH(OH)CO2)2·5H2O
Quantity 500 g
Residual Solvents Meets Requirements (USP)
Formula Weight 308.3
Percent Purity 98.0 to 101.0% (USP)
Grade USP
Physical Form Solid
Chemical Name or Material Calcium Lactate Pentahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.