Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Lactobionate Hydrate 97.0+%, TCI America™
SDP

Catalog No. L000625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L000625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L000625G Supplier TCI America Supplier No. L000625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5001-51-4
Molecular Formula C12H22O12
Molecular Weight (g/mol) 358.30
MDL Number MFCD00135895,MFCD10567434
InChI Key JYTUSYBCFIZPBE-AMTLMPIISA-N
Synonym calciumlactobionate
PubChem CID 131879797
IUPAC Name (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-{[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanoic acid
SMILES OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 754.65
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Lactobionate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.