Learn More
CPC Scientific H-Val-Lys-Phe-Gly-Val-Gly-Phe-Lys-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific CALP002B
Encompass
The peptide acts as a calmodulin agonist, interacts with the calmodulin EF-hand motif, the Ca2+ binding site. CALP-3 activates the phosphodiesterase in the absence of Ca2+ and inhibits cytotoxicity and apoptosis in a Ca2+ mediated manner.ONE-LETTER SEQUENCE: VKFGVGFKMOLECULAR FORMULA: C44H68N10O9MOLECULAR WEIGHT:881.09STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [261969-05-5], RESEARCH AREA: ApoptosisREFERENCES: M.Villain et al., J. Biol. Chem., 275, 2676 (2000); M.K.Manion et al., FASEB J., 14, 1297 (2000); R.Ten Broeke et al., FASEB J., 15, 1831 (2001); R.Ten Broeke et al., Eur. J. Pharmacol., 476, 151 (2003)
Catalog No. 50-210-8867