Learn More
CPC Scientific H-Val-Ala-Ile-Thr-Val-Leu-Val-Lys-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific CALP001A
Encompass
The peptide CALP-1 acts like Ca2+, interacts with the calmodulin-dependent proteins, like roponin superfamily, phosphodiesterase.ONE-LETTER SEQUENCE: VAITVLVKMOLECULAR FORMULA: C40H75N9O10MOLECULAR WEIGHT:842.09STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [145224-99-3], RESEARCH AREA: ApoptosisREFERENCES: J.Dillon et al., Proc. Natl. Acad. Sci. USA, 88, 9726 (1991); M.K.Manion et al., FASEB J., 14, 1297 (2000); R.Houtman et al., J. Immunol., 166, 861 (2001); R.Ten Broeke et al., Eur. J. Pharmacol., 476, 151 (2003)
Catalog No. 50-210-8864