Learn More
Hach Company Calcium/Magnesium Indicator Solution, 100 mL MDB
Supplier:  Hach Company 2241832
Encompass
For determination of calcium and magnesium hardness in water by the Calmagite Colorimetric method. Hach Method 8030. 100 mL Marked Dropping Bottle.
Catalog No. 50-199-5604