Learn More
Medchemexpress LLC Calcium mesoxalate trihydrate | 21085-60-9 | MFCD30541333 | 98.0% | 210.15 | C3H6CaO8 | 250 MG
SDP
Catalog No. 5000283992
Encompass_Preferred
Calcium mesoxalate trihydrate is a research-grade inorganic salt used in biochemical and cellular studies. It has reported biological activities including promotion of B cell proliferation and potential anti-diabetic effects, and is supplied as a high-purity, dry solid suitable for assay development and exploratory pharmacology.
  • Trihydrate form of calcium mesoxalate
  • Reported to promote B cell proliferation and show potential anti-diabetic activity
  • Provided as a high-purity solid (98.0%)
  • Molecular formula C3H6CaO8 and molecular weight 210.15
  • Available in small research quantities, including 250 mg
  • Suitable for biochemical assays and exploratory pharmacology
Catalog No. 50-002-83992 Supplier Medchemexpress LLC Supplier No. HYW540188A250MG
Calcium mesoxalate trihydrate is a research-grade inorganic salt used in biochemical and cellular studies. It has reported biological activities including promotion of B cell proliferation and potential anti-diabetic effects, and is supplied as a high-purity, dry solid suitable for assay development and exploratory pharmacology.
  • Trihydrate form of calcium mesoxalate
  • Reported to promote B cell proliferation and show potential anti-diabetic activity
  • Provided as a high-purity solid (98.0%)
  • Molecular formula C3H6CaO8 and molecular weight 210.15
  • Available in small research quantities, including 250 mg
  • Suitable for biochemical assays and exploratory pharmacology