Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Calcium Metaborate

Catalog No. 116181KGR Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-618-1KGR 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 11-618-1KGR Supplier Sigma Aldrich Supplier No. 116181KGR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Calcium metaborate can be used as an additive to improve the performance of nickel-metal hydride batteries.It is used as a precursor to synthesize calcium hexaboride (CaB6) by electro-reduction that can be used as high capacity anode for aqueous primary batteries.It can also be employed in the preparation of calcium-doped borate glass for thermoluminescent dosimeters (TLD).

Specifications

CAS 1315-04-4
Molecular Formula S5Sb2
Linear Formula Sb2S5
MDL Number MFCD00016321
Quantity 500 g
Synonym Calcium borate
Molecular Weight (g/mol) 403.85 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.