Learn More
Strem, An Ascensus Company CAS# 2556-53-8. 100g. Calcium methoxide, min. 90% (balance methanol). MFCD00058816
SDP
Catalog No. 5090114379
Encompass
CAS# 2556-53-8. 100g. Calcium methoxide, min. 90% (balance methanol). MFCD00058816. Molecular Weight: 102.15. Molecular Formula: Ca(OCH3)2. Color/form: off-white pwdr. Strem# 93-2062. http://www.strem.com/catalog/v/93-2062/
Catalog No. 50-901-14379 Supplier Strem, An Ascensus Company Supplier No. 932062100G
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
CAS# 2556-53-8. 100g. Calcium methoxide, min. 90% (balance methanol). MFCD00058816. Molecular Weight: 102.15. Molecular Formula: Ca(OCH3)2. Color/form: off-white pwdr. Strem# 93-2062. http://www.strem.com/catalog/v/93-2062/