Learn More
Medchemexpress LLC Calcium N5-methyltetrahydrofolate | 26560-38-3 | 99.85% | 497.52 | 1 MG
SDP
Catalog No. 5000386299
Encompass_Preferred
Calcium N5-methyltetrahydrofolate (NSC173328) is a derivative of Levomefolic acid that serves as a folic acid supplement. It can be used for research related to neural tube defects, infertility, non-Hodgkin's lymphoma, and endometriosis.
  • Effectively bypasses folate insufficiency
  • Uptake inhibited by Folic acid
Catalog No. 50-003-86299 Supplier Medchemexpress LLC Supplier No. HY175571MG
Calcium N5-methyltetrahydrofolate (NSC173328) is a derivative of Levomefolic acid that serves as a folic acid supplement. It can be used for research related to neural tube defects, infertility, non-Hodgkin's lymphoma, and endometriosis.
  • Effectively bypasses folate insufficiency
  • Uptake inhibited by Folic acid