Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calcium N5-methyltetrahydrofolate-d3 (racemic) | 68792-51-8 | MFCD22374090 | >98.0% | 500.54 g/mol | C20H20D3CaN7O6 | 1 MG
SDP

Catalog No. 5000284052
Encompass_Preferred

(rac)-Calcium N5-methyltetrahydrofolate-d3 is a deuterium-labeled, racemic calcium salt of 5-methyltetrahydrofolate supplied for research use. It is intended for use as a tracer and internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS. Handle and store according to recommended conditions to preserve isotopic integrity and stability.

  • Deuterium-labeled racemic 5-methyltetrahydrofolate calcium salt.
  • Suitable as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Supplied in a small research size for method development and validation.
  • Molecular formula C20H20D3CaN7O6; molecular weight 500.54 g/mol.
  • Storage: sealed at -20°C; in solvent stability -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-84052 Supplier Medchemexpress LLC Supplier No. HYW654270A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(rac)-Calcium N5-methyltetrahydrofolate-d3 is a deuterium-labeled, racemic calcium salt of 5-methyltetrahydrofolate supplied for research use. It is intended for use as a tracer and internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS. Handle and store according to recommended conditions to preserve isotopic integrity and stability.

  • Deuterium-labeled racemic 5-methyltetrahydrofolate calcium salt.
  • Suitable as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Supplied in a small research size for method development and validation.
  • Molecular formula C20H20D3CaN7O6; molecular weight 500.54 g/mol.
  • Storage: sealed at -20°C; in solvent stability -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.