Learn More
World Precision Instrument Calcium Neutral Ion Exchanger
SDP
Catalog No. 502538790
Encompass
Make micropipettes to record cellular concentrations
WPI’s Liquid Ion Exchangers (LIX), for use with the FD223A Electrometer, allow intracellular measurements to be made for cations (hydrogen, potassium and calcium).
Catalog No. 50-253-8790 Supplier World Precision Instrument Supplier No. IE200
Make micropipettes to record cellular concentrations
WPI’s Liquid Ion Exchangers (LIX), for use with the FD223A Electrometer, allow intracellular measurements to be made for cations (hydrogen, potassium and calcium).