Learn More
Bioworld Calcium Nitrate Tetrahydrate, 25 g
Catalog No. 50255671
Encompass_Preferred
It is a chemical used in the manufacture of fertilizers. Technical Information:Also known as Calcium nitrate 4-hydrateSensitivity: HygroscopicHazardous Materials Description:Calcium nitrateHazard class or Division: 5.1Identification Numbers: UN1454Packaging Group: III*Applies additional hazardous materials handling chargeAppearance: Crystalline or crystalline granules or flakesBoiling Point : 132°CDensity : 1.86 g/mL at 25 °CMelting Point: 42°C
Catalog No. 50-255-671 Supplier Bioworld Supplier No. 403000321
It is a chemical used in the manufacture of fertilizers. Technical Information:Also known as Calcium nitrate 4-hydrateSensitivity: HygroscopicHazardous Materials Description:Calcium nitrateHazard class or Division: 5.1Identification Numbers: UN1454Packaging Group: III*Applies additional hazardous materials handling chargeAppearance: Crystalline or crystalline granules or flakesBoiling Point : 132°CDensity : 1.86 g/mL at 25 °CMelting Point: 42°C